Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PAX6 Rabbit Polyclonal Antibody

Supplier: BioVision

3636R-100
10004-414EA 252.35 USD InStock
10004-414
Anti-PAX6 Rabbit Polyclonal Antibody
Antibodies
PAX genes encode nuclear transcription factors that may function as major controllers of developmental processes in both vertebrates and invertebrates. Mutations in murine PAX genes underlie three natural mouse alleles and several corresponding human syndromes (aniridia, foveal hypoplasia and Peters’ anomaly). Murine PAX genes have been shown to be proto-oncogenes. Furthermore, human PAX genes have recently been demonstrated to play an influential part in some common human cancers such as brain tumors and lymphomas. All PAX genes encode a DNA-binding domain termed the paired domain and in addition some also encode a second binding domain--the paired type homeobox. PAX6 is involved in the early development of the optical vesicle and has been shown to interact with Six3, another important visual development protein.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PAX6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty