Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FKBP12 Rabbit Polyclonal Antibody

Supplier: BioVision

3635-100
10004-408EA 252.35 USD InStock
10004-408
Anti-FKBP12 Rabbit Polyclonal Antibody
Antibodies
FKBP12 is a member of the family of soluble receptors capable of binding to one of two major immunosuppressant agents. FKBP12 also plays a role in intracellular calcium regulation by associating with three types of calcium release channel complexes, cardiac and skeletal ryanodine receptors and the inositol 1,4,5-trisphosphate receptor. In interactions with members of the TGF beta family type I receptors, FKBP12 has been shown to exert an inhibitory effect on the signaling pathway.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 2-10 µg/ml; Immunohistochemistry: 2-10 µg/ml

Type: Primary
Antigen: FKBP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty