Anti-VIM Rabbit Polyclonal Antibody

Supplier: BioVision
3634-100
10004-404EA 309.29 USD
10004-404
Anti-VIM Rabbit Polyclonal Antibody
Vimentin, a intermediate filament protein, is a general marker of cells originating in the mesenchyme. Vimentin is frequently co-expressed with other members of the intermediate filament family such as the cytokeratins, in neoplasm.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: VIM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR