Anti-SELPLG Rabbit Polyclonal Antibody

Supplier: BioVision
AB59738-100
10004-400EA 393.47 USD
10004-400
Anti-SELPLG Rabbit Polyclonal Antibody
Antibodies
Selectins are a family of carbohydrate-binding proteins involved in mediating cellular interactions with leukocytes. E-Selectin (also called ELMA-1) expressed on endothelial cells. L-Selectin (also called LECAM-1) is expressed on the majority of B and naïve T cells and on most monocytes, neurophils and eosinophils. P-Selectin (also called GMP-140) expressed on activated platelets and endothelial cells. Selectins interact with specific carbohydrates expressed by activated endothelial cells, monocytes, and neutrophils.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SELPLG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR