Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FN1 Rabbit Polyclonal Antibody

Supplier: BioVision

3630-100
10004-246EA 252.35 USD InStock
10004-246
Anti-FN1 Rabbit Polyclonal Antibody
Antibodies
Fibronectin is a glycoprotein synthesized in the liver for the circulating blood plasma form, and is synthesized by many mesenchymal cells, for the extracellular matrix form. It is composed of two similar, but not identical protein chains, which are linked by two disulfide linkages at the C-terminal end of the chains. The chains are composed of domains which have specific secondary structures linked together by regions which are especially susceptible to proteolytic action. For this reason, detection by immunoblot (western) may show considerable variability in the observed apparent molecular weights, which is predicated on the source of the fibronectin, and to what degree proteolysis has occurred. Bands approximately 225 kDa should be observed after SDS-PAGE when reducing and denaturing conditions are used

Recommended Dilutions: Western Blot: 0.5-2.0 µg/ml; Immunohistochemistry: 20-40 µg/ml

Type: Primary
Antigen: Fibronectin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty