Anti-HIST1H2BB Rabbit Polyclonal Antibody

Supplier: BioVision
3622-100
10005-704EA 309.39 USD
10005-704
Anti-HIST1H2BB Rabbit Polyclonal Antibody
The nucleosome is made up of four core histone proteins (H2A, H2B, H3 and H4) and is the primary building block of chromatin. The N-terminal tail of core histones undergoes different posttranscriptional modification including acetylation, phosphorylation and methylation. These modifications occur in response to cell signal stimuli and have a direct effect on gene expression.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 20-30 µg/ml

Type: Primary
Antigen: HIST1H2BB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR