Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSD17B4 Rabbit Polyclonal Antibody

Supplier: BioVision

3617-100
10004-220EA 252.35 USD InStock
10004-220
Anti-HSD17B4 Rabbit Polyclonal Antibody
Antibodies
HSD17B4 (17-beta-hydroxysteroid dehydrogenase 4) is a bifunctional enzyme that is involved in the peroxisomal beta-oxidation pathway for fatty acids. HSD17B4 catalyze the formation of 3-ketoacyl-CoA intermediates from both straight-chain and 2-methyl-branched-chain fatty acids.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: HSD17B4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty