Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TYH Rabbit Polyclonal Antibody

Supplier: BioVision

3612-100
10004-208EA 365.65 USD InStock
10004-208
Anti-TYH Rabbit Polyclonal Antibody
Antibodies
Tyrosine hydroxylase (TH) is the rate-limiting enzyme in the synthesis of the catecholamines Dopamine and Norepinephrine. TH antibodies can therefore be used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson’s disease and drμg abuse (Kish et al., 2001; Zhu et al., 2000; Zhu et al., 1999). TH antibodies can also be used to explore basic mechanisms of dopamine and norepinephrine signaling (Witkovsky et al., 2000; Salvatore et al., 2001; Dunkley et al., 2004). The activity of TH is also regulated by phosphorylation (Haycock et al., 1982; Haycock et al., 1992; Jedynak et al., 2002). Phospho-specific antibodies for the phosphorylation sites on TH can be used to great effect in studying this regulation and in identifying the cells in which TH phosphorylation occurs.

Recommended Dilutions: Western Blot: 1:1000; Iimmunofluorescence: 1:1000; Immunohistochemistry: 1:1000

Type: Primary
Antigen: TYH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mammalian, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty