Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDAC8 Rabbit Polyclonal Antibody

Supplier: BioVision

3608-100
10004-376EA 272.95 USD InStock
10004-376
Anti-HDAC8 Rabbit Polyclonal Antibody
Antibodies
Mammalian HDAC-8, isolated from human kidney, is a member of the histone deacetylase family. HDAC-8 shares homology with other HDACs in the family, but has different tissue distribution. HDAC-8 is localized to the nucleus and plays a role in the tissue development as well as cancer etiology.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: HDAC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty