Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDAC7 Rabbit Polyclonal Antibody

Supplier: BioVision

3607-100
10004-372EA 272.95 USD InStock
10004-372
Anti-HDAC7 Rabbit Polyclonal Antibody
Antibodies
Human HDAC7 is composed of 912 amino acid residues. Althoμgh HDAC7 is localized mostly to the cell nucleus, it is also found in the cytoplasm, sμggesting nucleo-cytoplasmic shuttling. The histone deacetylase activity of HDAC7 maps to a carboxy-terminal domain and is dependent on interaction with class I HDACs in the nucleus. HDAC7 catalyzes removal of acetyl-groups from acetyl-lysines of histones and promotes compaction of chromatin in these regions, leading to the inhibition of gene transcription.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 20-40 µg/ml; Immunohistochemistry: 20-40 µg/ml

Type: Primary
Antigen: HDAC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty