Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDAC6 Rabbit Polyclonal Antibody

Supplier: BioVision

3606-100
10004-368EA 305.96 USD InStock
10004-368
Anti-HDAC6 Rabbit Polyclonal Antibody
Antibodies
Human HDAC6 (1215 a.a. residues) possesses two separate putative catalytic domains, both of which are fully functional and contribute independently to the overall activity of HDAC6. A very potent NES is present at the amino-terminus of HDAC6, which was found to play an important role in regulating the shuttling of HDAC6 protein between cytoplasm and nucleus. The shuttling process may be a critical regulatory mechanism of HDAC6 function. HDAC6 may participate in coordinating expression of a group of genes involved in the remoldeling of chromatin during cell differentiation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 20 µg/ml

Type: Primary
Antigen: HDAC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty