Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDAC4 Rabbit Polyclonal Antibody

Supplier: BioVision

3604A-100
10004-360EA 252.35 USD InStock
10004-360
Anti-HDAC4 Rabbit Polyclonal Antibody
Antibodies
HDAC4 is a member of the class II mammalian histone deacetylases, which consists of 1084 amino acid residues. Its C-terminal sequence is highly similar to the deacetylase domain of yeast HDA1. HDAC4, unlike other deacetylases, shuttles between the nucleus and cytoplasm in a process involving active nuclear export. Association of HDAC4 with 14-3-3 results in sequestration of HDAC 4 protein in the cytoplasm. In the nucleus, HDAC4 associates with the myocyte enhancer factor MEF2A. Binding of HDAC4 to MEF2A results in the repression of MEF2A transcriptional activation. HDAC4 has also been shown to interact with other deacetylases such as HDAC3 as well as the corepressors NcoR and SMART.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: HDAC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chicken

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty