Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NFkB p50 Rabbit Polyclonal Antibody

Supplier: BioVision

3354R-100
10003-848EA 252.35 USD InStock
10003-848
Anti-NFkB p50 Rabbit Polyclonal Antibody
Antibodies
Nuclear factor kappa B (NFkB) was identified as a sequence specific transcriptional activator that binds to the intronic enhancer of kappa light chain gene in B lymphocytes. NFkB is a heterodimer that consists of a 50 kDa DNA binding subunit (p50) and a 65 kDa transactivation subunit (p65/RelA). Both of these subunits exhibit sequence homology to the protooncogene c-Rel. The p50 has an isoform called p49/p52, and both proteins are derived from the amino-terminal of precursor protein p105 and p100. The p50/p65 heterodimer remains in the cytosol in an inactive form as a complex with its inhibitor, IkB. Upon stimulation of cells by a wide variety of stimuli such as lipopolysaccharide (LPS), pro-inflammatory cytokines (IL-1 & TNF, etc.), and viral infection, IkB is phosphorylated and degraded by proteosome. The active NFkB heterodimer is translocated into the nucleus and induces gene expression.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: NFkB p50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty