Anti-CYCS Rabbit Polyclonal Antibody

Supplier: BioVision
3352-100
10003-840EA 285.95 USD
10003-840
Anti-CYCS Rabbit Polyclonal Antibody
Cytochrome c (m.w. 12,500) is an electron transport protein from mitochondria. It is released from mitochondria to cytoplasm during the early stages of apoptosis, prior to caspase activation, DNA fragmentation, and loss of membrane potential. The cytoplasmic cytochrome c is associated with Apaf-1 and caspase-9 to activate caspase-3 and other caspases.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CYCS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR