Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PCNA Rabbit Polyclonal Antibody

Supplier: BioVision

3350R-100
10003-836EA 288.4 USD InStock
10003-836
Anti-PCNA Rabbit Polyclonal Antibody
Antibodies
PCNA (Proliferating Cell Nuclear Antigen) is a 36 kDa protein also known as cyclin. In early S phase, PCNA has a very granular distribution and is absent from the nucleoli. At late S phase, PCNA is prominent in the nucleoli. There are two forms of PCNA protein present in cells, a soluble form sensitive to organic fixation and not involved in replication, and the other form is insoluble and is associated with ongoing DNA synthesis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PCNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty