Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TANK Rabbit Polyclonal Antibody

Supplier: BioVision

3348-100
10003-832EA 252.35 USD InStock
10003-832
Anti-TANK Rabbit Polyclonal Antibody
Antibodies
Members of TRAF family associate with activated TNF-R and CD40 and have been implicated to induce nuclear translocation of NFkB. TANK can bind to all three TRAFs and synergizing with TRAF2 to activate the NFkB signal cascade.

  • Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: TANK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty