Anti-BI-1 Rabbit Polyclonal Antibody

Supplier: BioVision
AB18852-100
10003-830EA 321.33 USD
10003-830
Anti-BI-1 Rabbit Polyclonal Antibody
Antibodies
BI-1 (Bax inhibitor-1), also designated TEGT for testis enhanced gene transcript, was identified as an additional inhibitor of apoptosis. BI-1 interacts with Bcl-2 and Bcl-xL, but not with Bax or Bak, to inhibit Bax induced apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BI-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR