Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FAF1 Rabbit Polyclonal Antibody

Supplier: BioVision

3346-100
10003-824EA 252.35 USD InStock
10003-824
Anti-FAF1 Rabbit Polyclonal Antibody
Antibodies
FAF1 (Fas-associated protein factor 1) specifically interacts with the cytoplasmic domain of wild type, but not mutated, Fas in both yeast and mammalian cells. When transiently expressed in T cells, FAF1 potentiates Fas-induced apoptosis. In contrast to TRADD, FADD, and RIP, FAF1 lacks a DDH (Death domain homologous region) and cannot induce apoptosis independent of Fas activation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in immunoprecipitation and Immunohistochemistry

Type: Primary
Antigen: FAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty