Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-POU2F1 Rabbit Polyclonal Antibody

Supplier: BioVision

3342-100
10003-818EA 252.35 USD InStock
10003-818
Anti-POU2F1 Rabbit Polyclonal Antibody
Antibodies
Members of Oct family of transcription factors specifically interact with Octamer motif ATGCAAAT, a regulatory element important for tissue- and cell-specific transcription as well as for transcription of a number of housekeeping genes. All of the members of the Oct family contain two highly conserved domains which are separated by 14-26 variable amino acids. These include the POU homeodomain and the POU-specific domain. Both are required for DNA binding and are involved in protein-protein interactions. Evidences indicate that regulation of Oct family transcription factors occurs at the level of phosphorylation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: POU2F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty