Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-P53DINP1 Rabbit Polyclonal Antibody

Supplier: BioVision

3340-100
10005-296EA 262.65 USD InStock
10005-296
Anti-P53DINP1 Rabbit Polyclonal Antibody
Antibodies
p53DINP1 (p53-dependent damage-inducible nuclear protein 1) encodes two proteins termed p53DINP1-α (27 kDa) and p53DINP1-β (18 kDa). p53DINP1 may regulate p53-dependent apoptosis throμgh phosphorylation at Ser46 and induction of p53AIP1. p53DINP1 is expressed in many tissues and induced by a variety of stress agents including UV stress, mutagenic stress, heat shock, and oxidative stress.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5-10 µg/ml)

Type: Primary
Antigen: P53DINP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty