Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PUMA/bbc3 Rabbit Polyclonal Antibody

Supplier: BioVision

3339R-100
10005-292EA 252.35 USD InStock
10005-292
Anti-PUMA/bbc3 Rabbit Polyclonal Antibody
Antibodies
PUMA (p53 upregulated modulator of apoptosis) is one of the pro-apoptotic Bcl-2 family members which are also transcriptional targets of p53. PUMA gene encodes two BH3 domain-containing proteins termed PUMA-α and PUMA-β. PUMA proteins bind Bcl-2, localize to the mitochondria, and induce cytochrome c release and apoptosis in response to p53. PUMA may be a direct mediator of p53-induced apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: PUMA/bbc3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty