Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BIRC7 Rabbit Polyclonal Antibody

Supplier: BioVision

3338-100
10003-814EA 272.95 USD InStock
10003-814
Anti-BIRC7 Rabbit Polyclonal Antibody
Antibodies
Livin is a novel member in the IAP (inhibitor of apoptosis) protein family. Livin contains a single baculoviral IAP repeat (BIR) domain and a RING finger domain and has two isoforms termed Livin-α and Livin-β. Transfection of Livin protected cells from apoptosis induced by FADD, BAX, RIP, RIP3 and DR6. Livin directly reacts with several caspases including Caspase-3, -7, and –9, and it inhibits the activation of caspase-9 induced by Apaf-1, cytochrome c and ATP.

Recommended Dilutions: Western Blot: 2-6 µg/ml)

Type: Primary
Antigen: BIRC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty