Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ENDOG Rabbit Polyclonal Antibody

Supplier: BioVision

3336R-100
10003-806EA 252.35 USD InStock
10003-806
Anti-ENDOG Rabbit Polyclonal Antibody
Antibodies
Endonuclease G (EndoG) is a mitochondrion-specific nuclease that specifically activated by apoptotic stimuli and translocates to the nucleus during apoptosis. In nucleus, EndoG induces nucleosomal fragmentation of DNA independent of caspase and DFF/CAD.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: ENDOG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty