Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FASLG Mouse Monoclonal Antibody [clone: I-6060]

Supplier: BioVision

3330-100
10003-794EA 303.85 USD InStock
10003-794
Anti-FASLG Mouse Monoclonal Antibody [clone: I-6060]
Antibodies
Fas (APO-1, CD95) is a 45 kDa cell surface protein that mediates apoptosis when cross-linked with agonistic anti-Fas antibodies or Fas ligand (FasL). FasL is a tumor necrosis factor (TNF)-related type II membrane protein and is expressed in various tissue and cells. Both Fas and FasL are thoμght to play an important role in the apoptosis processes that takes place during T cell development.

Recommended Dilutions: Western Blot: 1-4 µg/ml; immunoprecipitation, and Immunohistochemistry

Type: Primary
Antigen: Fas Ligand
Clonality: Monoclonal
Clone: I-6060
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty