Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IkBalpha Mouse Monoclonal Antibody [clone: 6A055]

Supplier: BioVision

3315-100
10003-790EA 303.85 USD InStock
10003-790
Anti-IkBalpha Mouse Monoclonal Antibody [clone: 6A055]
Antibodies
NF-kB is silenced in the cytoplasm by an inhibitory protein, IkB. IkB proteins are phosphorylated by IkB kinase complex consisting of at least three proteins, IKK1/α, IKK2/β, and IKK3/g. External stimuli such TNF-α or other cytokines results in phosphorylation and degradation of IkB releasing NF-kB dimers. NF-kB dimer subsequently translocates to the nucleus and activates target genes. Six members of IkB family members have been identified. One of the first gene induced following NF-kB activation is IkBα.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 1-4 µg/ml

Type: Primary
Antigen: IkBalpha
Clonality: Monoclonal
Clone: 6A055
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty