Anti-CIKS/Act1 Rabbit Polyclonal Antibody

Supplier: BioVision
3314-100
10003-788EA 309.39 USD
10003-788
Anti-CIKS/Act1 Rabbit Polyclonal Antibody
CIKS (for connection to IKK and SAPK/JNK), also called Act1 (for NF-kB activator 1), is a novel protein that associates with and activates IKK (IkB kinase). CIKS/Act1 also activates ATF (activating transcription factor) and AP-1 (activator protein 1) throμgh Jun kinase (JNK). CIKS/Act1 is ubiquitously expressed in human tissues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CIKS/Act1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR