Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DEDAF Rabbit Polyclonal Antibody

Supplier: BioVision

3313-100
10003-786EA 272.95 USD InStock
10003-786
Anti-DEDAF Rabbit Polyclonal Antibody
Antibodies
Cell death signal are transduced by death domain (DD), death effector domain (DED), and caspase recruitment domain (CARD) containing molecules. A novel protein that interacts with DED of caspase-8 and -10, and FADD was recently identified and designated DEDAF (for DED associated factor). DEDAF is identical to the transcriptional repressor RYBP. DEDAF interacts with FADD and aμgments the formation of CD95/FADD/caspase-8 complexes at the cytoplasm. DEDAF also interacts with DED-containing DNA biding protein (DEDD) in the nucleus, indicating it is involved in the regulation of both cytoplasmic and nuclear events of apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DEDAF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty