Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BCL2L1 Rabbit Polyclonal Antibody

Supplier: BioVision

3312-100
10003-782EA 293.55 USD InStock
10003-782
Anti-BCL2L1 Rabbit Polyclonal Antibody
Antibodies
Bcl-xL is a 30 kDa anti-apoptotic protein belonging to the Bcl-2 family. Bcl-xL prevents apoptosis throμgh two different mechanisms: by heterodimerization with an apoptotic protein to inhibit its apoptotic effect, and by its direct pore-forming effect on the outer-membrane of mitochondria to help maintain a normal membrane state under stress conditions. Bcl-xL can be cleaved by caspases upon apoptotic activation, which converts Bcl-xL from an antiapoptotic protein to a proapoptotic cleavage product. The cleavage product of Bcl-xL forms cytochrome c-releasing pores in the mitochondrial membrane to facilitate the apoptosis signaling pathway.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 20 µ/ml

Type: Primary
Antigen: BCL2L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty