Anti-RASA1 Rabbit Polyclonal Antibody

Supplier: BioVision
3311-100
10003-778EA 286.04 USD
10003-778
Anti-RASA1 Rabbit Polyclonal Antibody
The mammalian proto-oncogene Ras encodes 21 kDa proteins (p21Ras) which exists in either a physiologically quiescent GDP-binding state or a GTPbinding signal-emitting state. Interaction of p21Ras with GTPase activating protein (GAP) can increase hydrolysis of p21Ras-bound GTP by as much as 1000-fold. The product of the neurofibromatosis type 1 gene (NF1) has also been shown to exhibit p21Ras GAP activity.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunoprecipitation: 10-20 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: RASA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR