Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GSAP Rabbit Polyclonal Antibody

Supplier: BioVision

3308-100
10003-774EA 283.25 USD InStock
10003-774
Anti-GSAP Rabbit Polyclonal Antibody
Antibodies
GSAP, gamma-secretase activating protein interacts with gamma secretase which chops up the amyloid precursor protein to senile plaques in Alzheimer’s disease. GSAP was found to increase the production amyloid peptide. Immunohistochemistry to identify GSAP in brain can demonstrate its abundance in certain regions of damage, and is usually morphologically changed throughout the progression of Alzheimer's disease. GSAP can be used as a qualitative biomarker of Alzheimer's disease progression, and is regularly considered one of the best targets of therapies to diminish the effects of Alzheimer's disease in affected individuals.

Recommended Dilutions: Western Blot: 1-4 µg/ml;

Type: Primary
Antigen: GSAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty