Anti-CHK2 Rabbit Polyclonal Antibody

Supplier: BioVision
3305-100
10005-286EA 309.39 USD
10005-286
Anti-CHK2 Rabbit Polyclonal Antibody
Chk2 is the mammalian homologue of yeast Cds1/Rad53. In response to DNA damage, a checkpoint kinase ATM phosphorylates and activates Chk2, which in turn phosphorylates and activates p53. Chk2 also phosphorylates and activates BRCA1, the product of a tumor suppressor gene that is mutated in breast and ovarian cancer.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; immunofluorescence: 5-10 µg/ml

Type: Primary
Antigen: CHK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR