Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Smac Rabbit Polyclonal Antibody

Supplier: BioVision

3298-100
10003-768EA 252.35 USD InStock
10003-768
Anti-Smac Rabbit Polyclonal Antibody
Antibodies
Smac/DIABLO is a mitochondrial protein that is released along with cytochrome c during apoptosis and activates cytochrome c/Apaf-1/caspase-9 pathway. Smac/DIABLO binds to IAPs (Inhibitor of Apoptosis Proteins) and neutralizes the inhibitory effect of IAPs. Analysis of the structural basis of Smac/DIABLO has revealed that the N-terminal amino acids are required for Smac/DIABLO to bind to IAPs and activation of caspases.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5-20 µg/ml

Type: Primary
Antigen: Smac
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty