Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RIPK3 Rabbit Polyclonal Antibody

Supplier: BioVision

3293-100
10003-762EA 288.4 USD InStock
10003-762
Anti-RIPK3 Rabbit Polyclonal Antibody
Antibodies
Receptor interacting protein including RIP and RIP2/RICK mediate apoptosis induced by TNFR1 and Fas, two prototype members in the death receptor family. A novel member in the RIP kinase family was recently identified and designated RIP3. RIP3 contains N-terminal kinase domain but, unlike RIP or RIP2, lacks the c-terminal death or CARD domain. RIP3 binds to RIP and TNFR1, and mediates TNF-R1 mediated apoptosis, and attenuates RIP and TNFR1 induced NFkB activation. Overexpression of RIP3 induces apoptosis and NFkB activation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: RIPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty