Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BMP2 Rabbit Polyclonal Antibody

Supplier: BioVision

5672-100
10007-902EA 262.65 USD InStock
10007-902
Anti-BMP2 Rabbit Polyclonal Antibody
Antibodies
BMPs (bone morphogenetic proteins) belong to the TGF-beta superfamily of structurally related signaling proteins. Members of this superfamily are widely represented throμghout the animal kingdom and have been implicated in a variety of developmental processes. Proteins of the TGF-beta superfamily are disulfide-linked dimmers composed of two 12-15 kDa polypeptide chains. As implied by their name, BMPs initiate, promote and regulate bone development, growth, remodeling and repair. In addition to its osteogenic activity, BMP-2 plays an important role in cardiac morphogenesis. It is also expressed in a variety of tissues such as lung, spleen, brain, liver, prostate ovary and small intestine.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml

Type: Primary
Antigen: BMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty