Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GREM2 Rabbit Polyclonal Antibody

Supplier: BioVision

5657-100
10007-900EA 252.35 USD InStock
10007-900
Anti-GREM2 Rabbit Polyclonal Antibody
Antibodies
Gremlin (also known as Increased in High Glucose Protein 2, IHG-2, Down-regulated in Mos-transformed cells protein, Drm) functions as a bone morphogenetic protein (BMP) antagonist. It acts by binding to, and forming heterodimers with BMP-2, BMP-4, BMP-7, thus preventing them from interacting with their cell surface receptors.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Gremlin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty