Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RELM-beta Rabbit Polyclonal Antibody

Supplier: BioVision

5654-100
10007-898EA 252.35 USD InStock
10007-898
Anti-RELM-beta Rabbit Polyclonal Antibody
Antibodies
Human RELM beta (Resistin-like molecule beta/FIZZ2) is a new member to the family of adipocyte secreted proteins called adipocytokines. This family includes the RELM alpha, RELM beta and Resistin molecules. Interestingly, RELM beta and Resistin share similar characteristics such as an additional cysteine residue within the variable N-terminal region and are both homodimeric proteins. However, the RELM beta is expressed only in the gastrointestinal track; especially the colon, while the Resistin and RELM beta are secreted exclusively by adipocytes. Currently, the biological function of these proteins, as well as their molecular targets is largely unknown. Recombinant Human RELM beta is a disulfide-linked homodimer with a total molecular weight of 11.0 kDa, consisting of 90 amino acid residue chains.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: RELM-beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty