Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTHrP Rabbit Polyclonal Antibody

Supplier: BioVision

5652-100
10007-896EA 252.35 USD InStock
10007-896
Anti-PTHrP Rabbit Polyclonal Antibody
Antibodies
Parathyroid Hormone-like related Protein (PTHrP) is a member of the Parathyroid hormone family that is expressed in the mammary gland. It is known as a potent inhibitor of chondrocyte maturation. PTHrP is expressed by periarticular perichondrial cells and exerts its biological activity on PP-R expressing cells. During endochondral bone development PTHrP plays a major role in suppressing the onset of hypertrophic differentiation. It is also responsible for most cases of humoral hypercalcemia of malignancy. The antibody recognizes ~9.8 kDa of human PTHrP protein.

Recommended Dilutions: Western Blot: 0.5-4 ug/ml

Type: Primary
Antigen: PTHrP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty