Anti-TCRA Rabbit Polyclonal Antibody

Supplier: BioVision
AB18861-100
10007-888EA 747.76 USD
10007-888
Anti-TCRA Rabbit Polyclonal Antibody
Antibodies
TCR (T cell antigen receptor) recognizes foreign antigens and translates such recognition events into intracellular signals that elicit a change in the cell from a dormant to an activated state. TCR is a heterodimer composed of either α and β or γ and δ chains. The vast majority of circulating T cells (95%) express the α/β heterodimer while roμghly 2-5% express the γ/δ heterodimer. CD3 chains and the CD4 or CD8 coreceptors are also required for efficient signal transduction throμgh the TCR.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TCRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR