Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GAL7 Rabbit Polyclonal Antibody

Supplier: BioVision

5647-100
10007-886EA 252.35 USD InStock
10007-886
Anti-GAL7 Rabbit Polyclonal Antibody
Antibodies
Galectin-7 belongs to growing family of evolutionary conserved animal lectins. Galectins consists of beta-galactoside binding lectins that contain homologous carbohydrate recognition domains (CRDs). Galectin has the ability to induce apoptosis of activated T-cells and T-leukaemia cell lines. Other activities include cell differentiation and inhibition activity of CD45 protein phosphatase activity. Galectin also binds beta-galactoside as well as CD45, CD3 and CD4. Recombinant Human Galectin-7 is a 15.4 kDa protein consisting of 136 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Galectin-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty