Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VEGFB Rabbit Polyclonal Antibody

Supplier: BioVision

5641-100
10007-880EA 252.35 USD InStock
10007-880
Anti-VEGFB Rabbit Polyclonal Antibody
Antibodies
VEGF-B, a member of the VEGF family, is a potent growth and angiogenic cytokine. It promotes DNA synthesis in endothelial cells, regulates angiogenesis and vascular permeability, and inhibits apoptosis in certain smooth muscle cells and neurons. VEGF-B is expressed in all tissues except the liver. It forms cell surfaced-associated disulfide linked homodimers and can form heterodimers with VEGF-A. There are two known isoforms, formed by alternative splicing, which have been designated VEGF-B167 and VEGF-B186. Both forms have identical amino-terminal sequences encoding a “cysteine knot” like structural motif, but differ in their carboxyl-terminal domains. Both VEGF-B isoforms signal only throμgh the VEGFR1 receptor. Recombinant human VEGF-B is a 38.0 kDa disulfide-linked homodimeric protein consisting of two 167 amino acid polypeptide chains.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: VEGFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty