Anti-TGFBR2 Rabbit Polyclonal Antibody

Supplier: BioVision
5639R-100
10007-876EA 286.04 USD
10007-876
Anti-TGFBR2 Rabbit Polyclonal Antibody
TGF-β (Transforming growth factor-beta superfamily members are critical regulators of cell proliferation, differentiation, morphogenesis, and pathogenesis. TGF-beta receptor is a serine/threonine kinase receptor complex that consists of two distinct transmembrane proteins known as type I and type II receptors. In response to ligand binding, the type II receptors form a stable complex with the type I receptors allowing phosphorylation and thus activation of the type I receptor kinases.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TGFBR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR