Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TGF-beta Receptor I Rabbit Polyclonal Antibody

Supplier: BioVision

5636R-100
10007-872EA 252.35 USD InStock
10007-872
Anti-TGF-beta Receptor I Rabbit Polyclonal Antibody
Antibodies
TGF-β (Transforming growth factor-beta superfamily members are critical regulators of cell proliferation, differentiation, morphogenesis, and pathogenesis. TGF-beta receptor is a serine/threonine kinase receptor complex that consists of two distinct transmembrane proteins known as type I and type II receptors. In response to ligand binding, the type II receptors form a stable complex with the type I receptors allowing phosphorylation and thus activation of the type I receptor kinases.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: TGF-beta Receptor I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty