Anti-VEGFC Rabbit Polyclonal Antibody

Supplier: BioVision
5634-100
10007-870EA 286.04 USD
10007-870
Anti-VEGFC Rabbit Polyclonal Antibody
Vascular Endothelial Growth Factor-C (VEGF-C) is a 125 amino acid protein that plays an important role in angiogenesis and many other biological processes. Human VEGF-C exhibits about 85% homology with murine VEGF-C.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml;

Type: Primary
Antigen: VEGFC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR