Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VEGFC Rabbit Polyclonal Antibody

Supplier: BioVision

5633R-100
10007-868EA 252.35 USD InStock
10007-868
Anti-VEGFC Rabbit Polyclonal Antibody
Antibodies
Vascular Endothelial Growth Factor-C (VEGF-C) is a 125 amino acid protein that plays an important role in angiogenesis and many other biological processes. Human VEGF-C exhibits about 85% homology with murine VEGF-C.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: VEGFC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty