Anti-VEGFC Goat Polyclonal Antibody

Supplier: BioVision
5633-100
10007-866EA 344.42 USD
10007-866
Anti-VEGFC Goat Polyclonal Antibody
Vascular Endothelial Growth Factor-C (VEGF-C) is a 125 amino acid protein that plays an important role in angiogenesis and many other biological processes. Human VEGF-C exhibits about 85% homology with murine VEGF-C.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; Immunohistochemistry with both frozen and paraffin sections (5 µg/ml

Type: Primary
Antigen: VEGFC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR