Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RELM-alpha Rabbit Polyclonal Antibody

Supplier: BioVision

5631-100
10007-864EA 252.35 USD InStock
10007-864
Anti-RELM-alpha Rabbit Polyclonal Antibody
Antibodies
RELM-alpha belongs to a unique family of tissue-specific cytokines termed FIZZ (found in inflammatory zone) and RELM. The three known members of this family; Resistin, RELM-alpha and RELM-beta are 85-94 amino acid secreted proteins sharing a conserved C-terminal domain characterized by 10 cysteine residues with a unique spacing motif of C-X11-C-X8-C-X-C-X3-C-X10-C-X-C-X-C-X9-C-C. RELM-alpha and Resistin are secreted exclusively by adipocytes while RELM-beta is expressed in the epithelium of the colon and small bowel. The physiological role and molecular targets of RELM-alpha are still unknown. Murine RELM-alpha is a 10.0 kDa monomeric protein containing 88 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: RELM-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty