Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDAC8 Rabbit Polyclonal Antibody

Supplier: BioVision

5618-100
10007-862EA 252.35 USD InStock
10007-862
Anti-HDAC8 Rabbit Polyclonal Antibody
Antibodies
Human Histone Deacetylase 8 (HDAC8) is a member of the class I Histone Deacetylases. HDACs are important enzymes for the transcriptional regulation of gene expression in eukaryotic cells. HDACs catalyze the removal of acetyl groups from lysines near the N-termini of histones. Human HDACs have been implicated in a variety of human diseases such as cardiomyopathy, osteodystrophy, neurodegenerative disorders, aging and cancer. Expression of HDAC8 is restricted to cells showing smooth muscle differentiation in normal human tissue and is a novel marker of smooth muscle differentiation. Like other class I and II HDAC members, the activity of HDAC8 is sensitive to HDAC inhibitor Trichostatin A (TSA).

Recommended Dilutions: Western Blot: 1-5 µg/ml

Type: Primary
Antigen: HDAC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty