Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LIGHT Rabbit Polyclonal Antibody

Supplier: BioVision

5607-100
10007-860EA 272.95 USD InStock
10007-860
Anti-LIGHT Rabbit Polyclonal Antibody
Antibodies
LIGHT is a type II membrane protein belonging to the TNF family of membrane-anchored ligands. It is expressed in splenocytes, activated PBL, CD8+ tumor infiltrating lymphocytes, granulocytes and monocytes. LIGHT has the ability to activate NFkB, stimulate proliferation of lymphocytes, and induce apoptosis in certain human tumor cells. LIGHT binds to three receptors; the herpes virus entry mediator type A receptor (HEVM), LTbR and a decoy receptor (DcR3).

Recommended Dilutions: Western Blot: 0.5-1 µg/ml

Type: Primary
Antigen: LIGHT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty