Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IFN2B Rabbit Polyclonal Antibody

Supplier: BioVision

5595-100
10008-098EA 272.95 USD InStock
10008-098
Anti-IFN2B Rabbit Polyclonal Antibody
Antibodies
IFN-alpha 2b is a lymphoid factor that possesses potent anti-viral activity. Human IFN-a is a 19.3 kDa protein containing 165 amino acid residues.

Recommended Dilutions: Western Blot: 1-10 µg/ml

Type: Primary
Antigen: IFN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty