Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FGF20 Rabbit Polyclonal Antibody

Supplier: BioVision

5589-200
10008-096EA 252.35 USD InStock
10008-096
Anti-FGF20 Rabbit Polyclonal Antibody
Antibodies
Fibroblast Growth Factor-20 (FGF-20) is a secreted heparin binding growth factor belonging to the FGF family. FGF-20 signals throμgh the FGFR 2c and 3c and is expressed during limb and brian development. Human FGF-20 is a 23.2 kDa protein containing 209 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: FGF-20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty